Fume absorbers
Leverancier: LABCONCO
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3955421 3955420
LBCP3955421EA 0 EUR
LBCP3955421 LBCP3955420
Fume absorbers
Zuurkasten en behuizingen Afzuigkappen zonder afvoer
These fume adsorbers are for light duty histology cover slipping and staining areas. Activated carbon media adsorb organic odours and noxious vapors released by solvents like xylene and toluene while clean air is recirculated into the laboratory. These enclosures are low profile, completely portable and require no ductwork. They easily accommodate shallow countertops or those with overhanging cabinets.
  • Built-in, motorized impeller to provide face velocity up to 35 fpm
  • Glacier white, dry powder, epoxy-coated steel housing with perforated baffle for uniform airflow
  • Clear, one-piece, ³/₈" thick, shatter-resistant acrylic canopy
  • Pre-wired with rear-mounted auxiliary electrical receptacle and top-mounted blower switch with circuit breaker
  • Quiet operation at 58 to 63 dBA