Contact plates, gamma irridiated, triple-bagged, ICR (isolator, cleanroom)
Leverancier: MERCK LIFE SCIENCE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1462310200 1465520020 1461950200 1465520200 1461950020 1462310020 1465010200 1465380020 1465380200 1465390200
710-1893EA 1030 EUR
710-1893 HEIP1465520020 HEIP1461950200 HEIP1465520200 710-1891 710-1892 HEIP1465010200 HEIP1465380020 HEIP1465380200 HEIP1465390200
Contact plates, gamma irridiated, triple-bagged, ICR (isolator, cleanroom)
Media voor microbiologie Contactplaten voor microbiologie
ICR/ICRplus contact plates are triple-bagged and gamma irradiated to ensure the highest level of safety when monitoring surfaces in isolators and critical clean rooms for microorganisms.
  • Data matrix barcode on each plate for paperless and secure identification of individual plates
  • CLOSED and VENT closure options of ICRplus contact plates for safe transportation and variation of incubation conditions
  • Convenient room temperature stability allows storage at site of use