Settle plates, single-bagged
Leverancier: MERCK LIFE SCIENCE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1460040020 1460020020 1460020120 1460030120
HEIP1460040020EA 74 EUR
HEIP1460040020 HEIP1460020020 HEIP1460020120 HEIP1460030120
Settle plates, single-bagged
Media voor microbiologie Geprepareerde media voor microbiologie
A wide range of ready-to-use culture media for the environmental monitoring. 90 mm LI (Long Incubation) plates for the determination of the total cell count and for the test for specified microorganisms
  • Designed for the environmental monitoring in non critical areas