Rapid Translation System (RTS), Kits voor E. coli met hoge opbrengst
Leverancier: biotechrabbit
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BR1400101 BR1400102 BR1400201 BR1400301
733-2338EA 715 EUR
733-2338 733-2339 733-2341 733-2344
Rapid Translation System (RTS), Kits voor E. coli met hoge opbrengst
Eiwitexpressie Systemen voor eiwitexpressie
The Rapid Translation System (RTS) is a scalable in vitro protein expression platform that produces large amounts of protein for characterisation studies, functional assays, or structural analysis. RTS enables both transcription and translation utilising High-Yield (HY) and continuous-exchange, cell-free (CECF) technologies, with systems based on bacterial (E. coli) or eukaryotic (wheat germ) cell lysates. RTS offers the advantages of a cell-free protein expression system, whilst eliminating the need for laborious up and downstream steps typically associated with cell-based expression systems.
E. coli high yield kits
RTS 100 E. coli HY kit: Microgram-scale protein expression from multiple templates in parallel
Compatible with linear templates for fast screening without cloning
Convenient reaction set-up
Fast expression of proteins in parallel
Improved high yield biochemistry for up to 20 μg protein per reaction
RTS 500 ProteoMaster E. coli HY kit: Optimised medium scale cell-free expression of up to 6 mg protein
CECF technology ensures high yields
Enable expression of toxic proteins that cannot be produced in vivo
Express multiple proteins in parallel
Incorporate labelled methionine into synthesised proteins
RTS 9000 E. coli HY kit: Large scale, cell-free expression of up to 50 mg protein
CECF technology ensures high yields
Enable expression of toxic proteins that cannot be produced in vivo
Express multiple proteins in parallel
Incorporate labelled methionine into synthesised proteins