Culture media in bottles
Leverancier: Dr. Möller & Schmelz
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5201 5035 5080 5050 5030 5085 5101 5110 5005 5010 5038 5042 5040 5020 5021 5036 5060 5061 5062 5063 5081 5086 5090 5095 5100 5025
MOLL5201EA 59 EUR
MOLL5201 MOLL5035 MOLL5080 MOLL5050 MOLL5030 MOLL5085 MOLL5101 MOLL5110 MOLL5005 MOLL5010 MOLL5038 MOLL5042 MOLL5040 MOLL5020 MOLL5021 MOLL5036 MOLL5060 MOLL5061 MOLL5062 MOLL5063 MOLL5081 MOLL5086 MOLL5090 MOLL5095 MOLL5100 MOLL5025
Culture media in bottles
Media voor microbiologie Geprepareerde media voor microbiologie
These sterile ready-to-use media are filled in autoclavable and break-proof polycarbonate bottles (50 and 250 ml). The composition of the various types of media, comply with the formulations specified in the pertinent standards and regulations. Whereas the liquid media can be used immediately, the agar media first have to be remelted in a water bath at 90 to 100 °C. Once they have cooled down to around 50 °C you can pour agar plates in different sizes and thicknesses to meet your individual demands.
  • 4×250 ml in polycarbonate bottles