Antibiotic assay discs, differentiation discs (DD)
Leverancier: THERMO FISHER DIAGNOSTICS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
DD0001T DD0003T DD0004T DD0005T DD0013T DD0015T DD0008T
OXOIDD0001TEA 20.2 EUR
OXOIDD0001T OXOIDD0003T OXOIDD0004T OXOIDD0005T OXOIDD0013T OXOIDD0015T OXOIDD0008T
Antibiotic assay discs, differentiation discs (DD)
Testsystemen voor microbiologie Antibioticatesten
Used as a screening test prior to serological grouping.
  • Effect considerable economies in time, labour and materials
  • One cartridge contains 50 discs