Test tubes, standard ground joints, without stopper
Leverancier: Lenz Laborglas GmbH & CO.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
05700034 05700007 05700010 05700030 05700014 05700026 05700038 05700042
LENZ05700034EA 8.4 EUR
LENZ05700034 LENZ05700007 LENZ05700010 LENZ05700030 LENZ05700014 LENZ05700026 LENZ05700038 LENZ05700042
Test tubes, standard ground joints, without stopper
Buisjes Reageerbuisjes
DURAN®, borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
With standard ground joint, without stoppers.