Culture media, ready to use, in ampoules, Millipore®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MHA000P2T MHA00MRS2
MILXMHA000P2TEA 211 EUR
MILXMHA000P2T MILXMHA00MRS2
Culture media, ready to use, in ampoules, Millipore®
Media voor microbiologie Geprepareerde media voor microbiologie
Ready-to-use media solve numerous problems in microbiology laboratories where time, equipment and trained personnel are often in short supply. These media are available in convenient, single test 2 ml plastic ampoules, eliminating the manipulation of glass ampoules and avoiding the waste and spoilage associated with bulk packaged media. Manufacturing and testing of ampouled media is conducted in strict accordance with precise quality control procedures to ensure optimal growth and maximum shelf life. Reagent grade water is used to clean glassware and make media. Other factors that affect media stability and performance, such as finished product pH, entrained gases, light exposure and temperature are carefully analyzed and controlled during the manufacturing process.
  • Plastic ampoules
  • High quality
Order Now
Specificaties
Environmentally Preferable