Oxoid biochemical identification system (O.B.I.S.)
Leverancier: THERMO FISHER DIAGNOSTICS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ID0570M
ID0580M
ID0600M
OXOIID0570MEA
236
EUR
OXOIID0570M
OXOIID0580M
OXOIID0600M
Oxoid biochemical identification system (O.B.I.S.)
Testsystemen voor microbiologie
Enzymatische testen
The Oxoid Biochemical Identification System (O.B.I.S.) is a rapid colorimetric test for the determination of biochemical activity in microorganisms. It is a rapid screening method for differentiation of other species.
- Reduces the need for full biochemical identification of all suspect colonies