Coagulase plasma
Leverancier: DIFCO BD
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
240658 240827 240661
DIFC240658EA 141.7 EUR
DIFC240658 DIFC240827 DIFC240661
Coagulase plasma
Media voor microbiologie Additieven en supplementen voor microbiologie
Coagulase plasmas are used in qualitative procedures for the identification of Staphylococcus aureus.
  • Either citrated or EDTA lyophilized product configuration
Order Now
Specificaties
Environmentally Preferable