Coagulase plasma
Leverancier: DIFCO BD
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
240658
240827
240661
DIFC240658EA
141.7
EUR
DIFC240658
DIFC240827
DIFC240661
Coagulase plasma
Media voor microbiologie
Additieven en supplementen voor microbiologie
Coagulase plasmas are used in qualitative procedures for the identification of Staphylococcus aureus.
- Either citrated or EDTA lyophilized product configuration