Incubating orbital shakers, MaxQ™ HP
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
238000 238001 238008 238002
BARN238000EA 2290 EUR
BARN238000 BARN238001 BARN238008 BARN238002
Incubating orbital shakers, MaxQ™ HP
Schudapparaten Schudincubatoren
Max Q™ HP combines incubated or refrigerated temperature with an orbital shaker.
  • Triple counter-balanced mechanism
  • Microprocessor functions are calibrated through keypad
  • Clear acrylic lid withstands years of use without cracking or discoloration
  • Audible over and under-temperature alarms ensure sample protection
  • Innovative design ensures system protection in case of spills
Features a double walled insulated chamber which ensures temperature uniformity of ±0,3 °C. The spacious stainless steel chamber allows stacking of a 457×457 mm platform to hold six 2 L flasks. It is ideal for use in bacterial suspensions, solubility studies, extraction procedures, staining and destaining, hybridisations, diagnostic tests, washing procedures and protein expression.
Bestelinformatie: Supplied without platform or clamps which must be ordered separately.