Biochemical tests, DrySlide™
Leverancier: DIFCO BD
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
231746
231748
DIFC231746EA
417.75
EUR
DIFC231746
DIFC231748
Biochemical tests, DrySlide™
Testsystemen voor microbiologie
Antigeen-antilichaamtesten
Ag-AK, Immunoassay
The BD BBL DrySlide™ provides the alternative to biochemical testing. Reagent is dried on the slide, eliminating the need for reagent addition.
- Each slide is divided into four large segments, allowing for multiple testing on one slide
- Indole, results in 30 seconds
- Oxidase, results in 20 seconds