Eyewash ampoules dispenser, QuickRinse
Leverancier: Plum Safety
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
4632 5160
118-0550EA 38 EUR
118-0550 118-0481
Eyewash ampoules dispenser, QuickRinse
Eerste-hulpbenodigdheden Oogdouches
This wall-mountable dispenser contains 10 (2×5) ampoules with 20 ml of 0,9 % sterile sodium chloride eyewash solution.
  • Quick, easy and hygienic help for rinsing dust and dirt out of the eyes
  • Compact station is easy to mount on the wall
  • Station is refillable
Shelf life: 3 years (when unopened)
W×H: 230×135 mm