Tissue grinders, Pellet Pestle®, KIMBLE®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SP749515-0000 SP749516-0500
SCERSP749515-0000EA 212 EUR
SCERSP749515-0000 SCERSP749516-0500
Tissue grinders, Pellet Pestle®, KIMBLE®
Homogenisatoren Potterbuizen
Pellet pestles with a stainless steel shaft/CTFE tips.
  • Reusable pellet pestles for microcentrifuge and centrifuge tubes