Electrode Plug Cable Combinations
Leverancier: SI Analytics
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
285124613 285124654 285124708
SCOI285124613EA 42.4 EUR
SCOI285124613 SCOI285124654 SCOI285124708
Electrode Plug Cable Combinations
Kabels
Electrode cables along with electrode socket, 1 m two core cable up to 80 °C, for resistance thermometer and 2-pole-conductivity cell without temperature sensor. Available with different cable length.
Order Now
Specificaties
Environmentally Preferable