Bottles, narrow neck, HDPE, with screw cap
Leverancier: LAMAPLAST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
351 352 353 356 354
215-4121EA 2 EUR
215-4121 215-4122 215-4123 215-4126 215-4124
Bottles, narrow neck, HDPE, with screw cap
Flessen Kunststof flessen
HDPE.
  • Leakproof
  • Suitable for food stuffs
  • Screw cap with opening-off seal
Ideal for storage of very fluid liquids. Complete emptying is guranteed by the inclination of the shoulders.