Suction filters, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
F765-16 F765-56 F765-08 F765-48
BOHLF765-16EA 139 EUR
BOHLF765-16 BOHLF765-56 BOHLF765-08 BOHLF765-48
Suction filters, BOLA
Filters Filters voor vloeistofverwerking
These PTFE suction filters consist of a receiver with a female thread and an easily exchangeable frit. They are ideal prefilters in front of pump systems for protecting gaskets, pistons or column packings against premature wearing due to contaminations.
Good chemical resistance
Temperature resistant from −200 to +250 °C