Bulk packing material for preparative HPLC columns, SepTech™ C18 reverse phase media
Leverancier: VARIAN
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
A80600100G
A8060001KG
A8150001KG
A81500100G
VARIA80600100GEA
1020
EUR
VARIA80600100G
VARIA8060001KG
VARIA8150001KG
VARIA81500100G
Bulk packing material for preparative HPLC columns, SepTech™ C18 reverse phase media
Sorbenten voor chromatografie
Sorbenten voor HPLC
SepTech™ C18 bulk media, designed for high performance separations at high capacity. The two pore sizes, 60 Å and 150 Å, have been optimised to give high capacity for small molecules and larger natural molecules and peptides, respectively.