Flatbed precast gels for IEF, SERVALYT™ PRECOTES™ CSF
Leverancier: Serva
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
42800.01 42801.01 42866.02 42867.02 42874.02 42875.02 42878.02 42919.03 42965.03 42967.02 42974.02 42978.02
SERA42800.01EA 436 EUR
SERA42800.01 SERA42801.01 SERA42866.02 SERA42867.02 SERA42874.02 SERA42875.02 SERA42878.02 SERA42919.03 SERA42965.03 SERA42967.02 SERA42974.02 SERA42978.02
Flatbed precast gels for IEF, SERVALYT™ PRECOTES™ CSF
Gel voor elektroforese Voorgegoten gels
For cerebrospinal fluid (CSF) analysis by isoelectric focusing.
Leveringsinformatie: SERVALYT™ PRECOTES™ CSF Kit contains:
5× SERVALYT™ PRECOTES™ CSF gels, 245×125 mm, 300 µm
Anode and cathode buffer solutions
Applicator strips and electrode wicks
Optimised protocol for silver staining