Vial racks, for Cetac autosampler
Leverancier: PerkinElmer
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
N0774085 N0777154 N0777298
PERCN0774085EA 554 EUR
PERCN0774085 PERCN0777154 PERCN0777298
Vial racks, for Cetac autosampler
Rekken Rekken voor chromatografievials
These polypropylene racks accommodate 40 headspace auto-sampler vials.
  • Autoclavable and stackable
  • Good chemical and temperature resistance
  • Features end plates for labeling and numbering for tube identification