Flip tube A
Leverancier: HAMILTON ROBOTICS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
235693 235454
HAMR235693EA 555 EUR
HAMR235693 HAMR235454
Flip tube A
Buisjes Buisjes voor robotsystemen
The consumable flip tube® A is especially designed to automate the opening / closing of the lid by a controlled automated process, that applies a stepwise gentle pressure on the rear end of the hinged lid, enabling a fully walkaway automated process.
  • Made from highly transparent polypropylene for easy optical control
  • Contamination-free opening manually or by FlipTube® tool
  • Precise lid sealing to minimize sample evaporation
  • Pierceable lid and easy marking due to large frosted lid and surface on the side
  • Available in 'standard quality', 'biological purity tested' and 'biological purity PLUS' quality grade for guaranteed purity