Bottle top dispenser, Calibrex™ organo 525
Leverancier: Socorex Isba
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
525.025 525.050 525.100 525.025FC 525.050FC 525.100FC 525.001 525.001FC 525.002.5 525.002.5FC 525.005 525.005FC 525.010 525.010FC
613-5976EA 648 EUR
613-5976 613-5978 613-5980 613-5977 613-5979 613-5981 613-5968 613-5969 613-5970 613-5971 613-5972 613-5973 613-5974 613-5975
Bottle top dispenser, Calibrex™ organo 525
Flesdispensers
Robust dosing instruments with excellent chemical resistance, high performance and simplified maintenance for safe and reproducible distribution ranging from 0,1 to 100 ml.
  • Ground glass or ceramic plunger for organic and non-crystallizing acid and base solutions
  • QR coded chemical compatibility
  • Integrated calibration key
  • Quick disassembling, no tool required
  • Autoclavable at 121 °C fully assembled
Flow control stopcok available, for priming without reagent loss or contamination, with locking position for safe dispenser transportation. Screw-locking stopper.
Certificaten: Individual QC certificate according ISO 8655.
Leveringsinformatie: Supplied with 45 mm base thread, 120 mm delivery jet and stopper, 350 mm feed tube, alternative screw-type volume setting button, three bottle neck adapters (32, 38 and 40 mm), QC certificate and operating instructions. Optional extended delivery jet (150 mm) available as accessory.