Tubings, insulated
Leverancier: LAUDA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
LZM052 LZM053 LZM054 LZM055
LAUDLZM052EA 291 EUR
LAUDLZM052 LAUDLZM053 LAUDLZM054 LAUDLZM055
Tubings, insulated
Slangen Transfer Tubing
Metal hoses with insulation for connections with thread M16×1.
  • Temperature resistance: −90 to +150 °C.