Cylindrical bend testers
Leverancier: Industrial Physics
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SP1820 SP1822
SIMESP1820EA 2180 EUR
SIMESP1820 SIMESP1822
Cylindrical bend testers
Coatinglaag testers
These cylindrical bend testers are used to determine the elasticity, adhesion and elongation of paint on metal.
  • Sturdy conception in anodised aluminium and stainless steel
  • Ergonomic clamping device for test panels
  • Large knob on bending arm for smooth bending
The test panel is bent over the cylindrical mandrel and then checked for damages.
Certificaten: ISO 1519 (or ASTM D522)
Leveringsinformatie: Supplied with a desktop holder and mandrels.