Dropping pipettes
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SBD050 SBD100 SBD250
BIBBSBD050EA 102 EUR
BIBBSBD050 BIBBSBD100 BIBBSBD250
Dropping pipettes
Pipetten Pasteur-/transferpipetten
Soda-lime glass.
  • With integral ground glass cone on pipette
  • Supplied with rubber teats
  • Bottles available separately