Septa caps
Leverancier: EP SCIENTIFIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
S297-5300 297-2414 24-414/WS-3M 297-2400UB
EPSCS297-5300EA 219 EUR
EPSCS297-5300 EPSC297-2414 EPSC24-414/WS-3M EPSC297-2400UB
Septa caps
Sluitingen Flesdoppen
PP, white.
Septa caps fit VOA vials, bottles and jars. The septum is permanently bonded to the cap and processed to meet EPA performance based specifications for volatile organic analysis.