Transport and Storage Containers, ITB
Leverancier: BEHR
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
B00179010 B00179009
331-3221EA 690 EUR
331-3221 331-3222
Transport and Storage Containers, ITB
Dozen Transportdozen
These transport containers are suitable for storage and transport of 6 or 8 DIN-sampling bottles. They are ideal for the storage of light sensitive samples. The cooling element in the lid ensures to maintain cooling inside the sample container. The insulated cladding eliminates the influences of temperature change in the samples.
  • Stackable
  • Space saving