Baths and lids, digital, stainless steel
Leverancier: witeg Labortechnik
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
DH.WB000111 DH.WB000122 DH.WB000106
WITGDH.WB000111EA 447 EUR
WITGDH.WB000111 WITGDH.WB000122 WITGDH.WB000106
Baths and lids, digital, stainless steel
Baden Waterbaden, ongeroerd
These stainless steel baths are ideal for biotechnology, clinical, environmental, medical, petroleum, pharmaceutical or industrial applications.
  • Stainless steel lid prevents evaporation and keeps temperature constant
  • Durability and high thermal efficiency
  • Digital fuzzy controller with jog - shuttle switch
  • Digital LCD display