Bottle racks
Leverancier: MERCK LIFE SCIENCE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1465250001 1465260001
HEIP1465250001EA 204 EUR
HEIP1465250001 HEIP1465260001
Bottle racks
Rekken Flessenrekken
Racks for media bottles, made of high quality stainless steel, electrolytic polished.
  • Autoclavable
  • Easy to clean
  • Good chemical and temperature resistance
Safe and convenient transport of bottles. Suitable for isolators and clean rooms.