Septa for open top caps
Leverancier: EP SCIENTIFIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
288-0022BP S24-400-S2 288-0022 288-7222
EPSC288-0022BPEA 2250 EUR
EPSC288-0022BP EPSCS24-400-S2 EPSC288-0022 EPSC288-7222
Septa for open top caps
Tussenschoten
These silicone septa with PTFE coating are suitable for VOA vials and bottles with 24-414 open-top cap.
  • Processed to meet EPA Performance Based Specifications for Volatile Organic Analysis