Anti-Glypican 4 Rabbit Polyclonal Antibody

Leverancier: United States Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

USBIG8235-52D-20EA 394 EUR
USBIG8235-52D-20 USBIG8235-52D-100
Anti-Glypican 4 Rabbit Polyclonal Antibody
Antilichamen
Recommended dilution: Western Blot: 1:1000-1:2000; Immunochemistry: 1:1000

Type: Primary
Antigen: Glypican 4
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human,
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR