Anti-Insulin-like Growth Factor I Rabbit Polyclonal Antibody

Leverancier: United States Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

USBII7661-01S-20EA 371 EUR
USBII7661-01S-20 USBII7661-01S-100
Anti-Insulin-like Growth Factor I Rabbit Polyclonal Antibody
Antilichamen
Recommended dilution: Western Blot: 1:1000-1:2000

Type: Primary
Antigen: Insulin-like Growth Factor I
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human,
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR