Anti-Parathyroid Hormone Mouse Monoclonal Antibody [clone: B1/70
]
Leverancier: US Biological
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
P3109-10E-10 P3109-10E-100
USBIP3109-10E-10EA 371 EUR
USBIP3109-10E-10 USBIP3109-10E-100
Anti-Parathyroid Hormone Mouse Monoclonal Antibody [clone: B1/70
]
Antilichamen
Recommended dilution: ELISA: 1 ?g/mlRadio Immunoassay: 25ng/ml; Immunochemistry: 2?g/ml
Type: Primary
Antigen: Parathyroid Hormone
Clonality: Monoclonal
Clone: B1/70
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human,