Anti-IGF1Rb Rabbit Monoclonal Antibody [clone: 10E33]
Leverancier: United States Biological
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
USBII7661-21M2-100EA 681 EUR
USBII7661-21M2-100
Anti-IGF1Rb Rabbit Monoclonal Antibody [clone: 10E33]
Antilichamen
Recommended dilution: Western Blot: 1:1000
Type: Primary
Antigen: IGF1Rb
Clonality: Monoclonal
Clone: 10E33
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse,