Anti-Phosphatidylinositol Phosphate Rabbit Polyclonal Antibody
Leverancier: US Biological
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
141851 141851-100
USBI141851EA 364 EUR
USBI141851 USBI141851-100
Anti-Phosphatidylinositol Phosphate Rabbit Polyclonal Antibody
Antilichamen
Recommended dilution: ELISA: 1:100-200; Western Blot: 1:100-400; Immunochemistry: 1:100-500
Type: Primary
Antigen: Phosphatidylinositol Phosphate
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human,