Anti-HSPD1 Mouse Monoclonal Antibody (Unconjugated) [clone: LK2]
Leverancier: Rockland Immunochemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ROCK200-301-F61EA 798 EUR
ROCK200-301-F61
Anti-HSPD1 Mouse Monoclonal Antibody (Unconjugated) [clone: LK2]
Antilichamen
Anti-Hsp60 Antibody is suitable for WB, Flow Cytometry, and IHC. Expect a band approximately ~60kDa Protein corresponding to the molecular mass of Hsp60 on SDS PAGE immunoblots.
Recommended Dilutions: Western Blot: 0.25 µg/ml;
Type: Primary
Antigen: HSPD1
Clonality: Monoclonal
Clone: LK2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Bovine, Dog, Chicken, Guinea Pig, Hamster, Monkey, Pig, Rabbit, Spinach, E. coli, H. Pylori, S. Typhimurium, T. Spiralis , Yeast, White Fly