Anti-ALPI Goat Polyclonal Antibody
Leverancier: Rockland Immunochemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
200-1135S 200-1135-0100
ROCK200-1135SEA 226 EUR
ROCK200-1135S ROCK200-1135-0100
Anti-ALPI Goat Polyclonal Antibody
Antilichamen
Recommended Dilutions: ELISA: 1:4000 - 1:20000; Western Blot: 1:500 - 1:2000; Other Dilution: User Optimized
Type: Primary
Antigen: ALPI
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine