Anti-PCNA Mouse Monoclonal Antibody [clone: IPO-38]

Leverancier: Cayman Chemical
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CAYM10004805-1EA 468 EUR
CAYM10004805-1
Anti-PCNA Mouse Monoclonal Antibody [clone: IPO-38]
Antilichamen
Type: Primary
Antigen: PCNA
Clonality: Monoclonal
Clone: IPO-38
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: multiple species
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR