Anti-CD36 Mouse Monoclonal Antibody [clone: JC63.1]
Leverancier: Cayman Chemical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
10009893-100 10009893-500
CAYM10009893-100EA 478 EUR
CAYM10009893-100 CAYM10009893-500
Anti-CD36 Mouse Monoclonal Antibody [clone: JC63.1]
Antilichamen
Type: Primary
Antigen: CD36
Clonality: Monoclonal
Clone: JC63.1
Conjugation: unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgA
Reactivity: Human, Mouse, Rat