Anti-Hsf2 Rat Monoclonal Antibody [clone: 3E2]
Leverancier: Cayman Chemical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
10011434-100 10011434-25
CAYM10011434-100EA 713 EUR
CAYM10011434-100 CAYM10011434-25
Anti-Hsf2 Rat Monoclonal Antibody [clone: 3E2]
Antilichamen
Type: Primary
Antigen: Hsf2
Clonality: Monoclonal
Clone: 3E2
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG
Reactivity: Bovine, Dog, Guinea Pig, Hamster, Human, Monkey, Mouse, Sheep, Pig, Rabbit, Rat