Anti-Nampt Mouse Monoclonal Antibody [clone: OMNI 379]
Leverancier: Cayman Chemical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
10813-50 10813-100
CAYM10813-50EA 423 EUR
CAYM10813-50 CAYM10813-100
Anti-Nampt Mouse Monoclonal Antibody [clone: OMNI 379]
Antilichamen
Type: Primary
Antigen: Nampt
Clonality: Monoclonal
Clone: OMNI 379
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human, Mouse, Rat