Anti-Vtg Rabbit Polyclonal Antibody [clone: CS-1]
Leverancier: Cayman Chemical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
170130-100 170130-500
CAYM170130-100EA 520 EUR
CAYM170130-100 CAYM170130-500
Anti-Vtg Rabbit Polyclonal Antibody [clone: CS-1]
Antilichamen
Type: Primary
Antigen: Vtg
Clonality: Polyclonal
Clone: CS-1
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Atlantic Cod, common Carp, Rainbow Trout, Tilapia, Turbot