Spuitfilters, analytische filtratie, Nalgene®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
171-0020 171-0045 176-0020 176-0045 728-2045 721-1320 721-1345 722-2000 724-2020 724-2045 724-2080 727-2020 727-2045 728-2020
514-0455EA 248 EUR
514-0455 513-1712 513-1761 513-1762 516-0950 516-0931 516-0932 516-0933 516-0940 516-0941 516-0942 516-0947 516-0948 516-0949
Spuitfilters, analytische filtratie, Nalgene®
Filters Spuitfilters
These syringe filters are ideal for filtration, clarification or particle removal from buffers, reagents, DMSO, HPLC/GC solvents.
  • Sterile filters are certified sterile, non-cytotoxic and non-pyrogenic
  • Accommodate sample volumes from 10 to 100 ml
SFCA membrane; PP housing: Use for immunological samples and biological fluids. Low protein binding, lower extractables than standard CA.
CA membrane; PP housing: Ideal for buffers, aqueous reagents and solutions, including EIA and ELISA samples. Some protein binding and extractables. For sterile filtrationMore Product Information