Anti-Foxp3 Rat Antibody (APC (Allophycocyanine)) [clone: MF23]
Leverancier: Tonbo Biosciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
20-0191-U025 20-0191-U100
20-0191-U025EA 111 EUR
20-0191-U025 20-0191-U100
Anti-Foxp3 Rat Antibody (APC (Allophycocyanine)) [clone: MF23]
Antilichamen
The MF23 antibody reacts with mouse Foxp3, a 50-55 kDa transcription factor which is a central regulator of T cell activity and is critical for the development and function of regulatory T cells (Tregs). Foxp3 is a member of the forkhead box or winged helix family of transcription factors and is expressed at constitutively high levels in Treg cells. Forced expression of Foxp3 in conventional T cells results in the upregulation of Treg associated molecules such as CD25, CTLA-4, and GITR and is sufficient to impart suppressive functional activity to these cells.
Type: Primary
Antigen: Foxp3
Clonality:
Clone: MF23
Conjugation: APC (Allophycocyanine)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse