Spectrophotometer cuvettes
Leverancier: Lovibond Tintometer
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
601040 601050
TINT601040EA 53.4 EUR
TINT601040 TINT601050
Spectrophotometer cuvettes
Cuvetten Cuvetten voor spectrofotometers
These optical glass spectrophotometer cuvettes can be used in PFXi/PFX instruments.
These cuvettes can be used in PFXi/PFX instruments when the 12,5 mm holder is used.