Bags, zip seal, black opaque, writing area, Minigrip®
Leverancier: Flexico
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ITM025752 ITM025753 ITM025751
129-2362EA 304 EUR
129-2362 129-2363 129-2361
Bags, zip seal, black opaque, writing area, Minigrip®
Zakken Monsterzakken
PE bags, thickness 60 µm.
  • Confidentiality of content due to opacity to light
  • 3 white write-on panels
  • Hanging hole
  • Resealable, reusable, 100% recyclable
Made of polyethylene (LDPE). Zip seal designed for easy opening and high speed packing. The Minigrip® bag provides an excellent mechanical resistance and high temperature resistance (from –40 up to +60 °C). NOT suitable for food contact.