Bouffant caps, Hopen®
Leverancier: Medicom
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
70.013 70.012 70.104 70.061 70.011S 70.011 70.102 70.062 70.101 70.103
HOPE70.013EA 31.1 EUR
HOPE70.013 HOPE70.012 HOPE70.104 HOPE70.061 113-8336 HOPE70.011 HOPE70.102 HOPE70.062 HOPE70.101 HOPE70.103
Bouffant caps, Hopen®
Wegwerpkleding
These disposable bouffant caps made of PP are ideal for use in hygiene and food industries.
  • Cover the hair as required by HACCP standards
  • Elasticated for perfect fit
  • Available in different colours and different packaging options
Weight: 12 g/m²
Order Now
Specificaties
Environmentally Preferable
Geslacht
Kledingstuk toepassing Food