SPE tubes, Supelclean™ ENVI-18, Supelco®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
57062 57063 57064 57114 57137 57138 57219
57062.EA 236 EUR
57062. 57063. 57064. 57114. 57137. 57138. 57219.
SPE tubes, Supelclean™ ENVI-18, Supelco®
Kolommen voor chromatografie
This Reversed phase Supelclean™ ENVI-18 are suitable for excellent for cleaning, extracting and concentrating pollutants from aqueous environmental sample (drinking, ground, waste water). Also suitable for extracting herbicides, fungicides and pesticides from waste material.
  • Polymerically bonded, octadecyl, endcapped
  • Higher carbon loading (17%)
  • Increased binding capacities and higher recoveries
  • Resistance to extreme pH conditions