White light box
Leverancier: Edvotek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
733-2003EA 213 EUR
733-2003
White light box
Lampen Lichtbakken
The Edvotek White Light Box features a spacious 25×25 cm viewing area illuminated by long life LEDs and is housed in a slim aluminium body. It is designed to safely enhance the visualisation of DNA stained with FlashBlue™, proteins stained with Coomassie Blue and autoradiograms.
Power: 8 W
Dual Voltage: 110/220 V, 50/60 Hz
Dimensions: 345×248 mm